Assay Details
| Gene Symbol: MUC5AC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304359.1 | NP_001291288.1 | 1619 | 76 | ||
| GenBank mRNA | AF015521.1 | - | 13 - 14 | 1619 | 76 | |
| AF043909.1 | - | 13 - 14 | 1610 | 76 | ||
| AJ298317.1 | - | 13 - 14 | 1610 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145288.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145288.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145288.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145288.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145288.hg.1 | TC1100006508.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | ENST00000621226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006508.hg.1 | NM_001304359.1 | Refseq |

MUC5AC
Genomic Map