| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016526.hg.1 |
ENST00000623247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016526.hg.1 |
ENST00000623247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016526.hg.1 |
ENST00000623247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016526.hg.1 |
ENST00000623247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016527.hg.1 |
ENST00000445098.2 |
Ensembl |