Assay Details
| Gene Symbol: LINC00083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003086.1 | XP_016858575.1 | 8 - 9 | 1250 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685282 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626130 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685245 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15147129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15147145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15147132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626128 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626111 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626096 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626171 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20685230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626131 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00626160 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 22795 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 375231 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 247327 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 49715 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 95520 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 96939 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30385103 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00200587 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00200584 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00200581 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7922593 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7922591 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7922595 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7922594 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7922592 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7922590 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1763975 | AK125993.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1763975 | AK125993.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1763975 | AK125993.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1763975 | AK125993.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1763975 | AK125993.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1763975 | AK125993.1 | human_transcriptome |

LINC00083
Genomic Map