| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021932.hg.1 |
TC09001719.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021932.hg.1 |
ENST00000298466.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021932.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021932.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021932.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181660.hg.1 |
TC0900011950.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |