Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142645.1 | NP_001136117.1 | 5 - 6 | 701 | 80 | |
| XM_005246212.3 | XP_005246269.1 | 5 - 6 | 855 | 80 | ||
| XM_005246213.3 | XP_005246270.1 | 5 - 6 | 540 | 80 | ||
| XM_011510458.2 | XP_011508760.1 | 5 - 6 | 681 | 80 | ||
| XM_011510459.2 | XP_011508761.1 | 5 - 6 | 886 | 80 | ||
| XM_017003098.1 | XP_016858587.1 | 5 - 6 | 688 | 80 | ||
| XM_017003099.1 | XP_016858588.1 | 5 - 6 | 688 | 80 | ||
| XM_017003100.1 | XP_016858589.1 | 5 - 6 | 688 | 80 | ||
| GenBank mRNA | AK296987.1 | - | 5 - 6 | 649 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002620.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02041979.hg.1 | TC02002620.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041979.hg.1 | ENST00000409150.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041979.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041978.hg.1 | TC02002620.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041978.hg.1 | ENST00000409150.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041978.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211578.hg.1 | TC0200015231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211579.hg.1 | TC0200015231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |

NEMP2
Genomic Map