Assay Details
| Gene Symbol: GGTA1P | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF378121.1 | - | LIT2093 alpha1,3GTP1 |
7 - 8 | 818 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stomatin | STOM |
BND7, EPB7, EPB72 |
-74.7kb | Sense | Chr. 9 - 121338988 - 121370304 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000373793.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000481799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000488429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | ENST00000495328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_003191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011407.hg.1 | NR_045211.1 | Refseq |

GGTA1P
Genomic Map