Assay Details
Gene Symbol: TMEM8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080483.2 | NP_001073952.1 | 2 - 3 | 354 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001692.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09021719.hg.1 | TC09001692.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09021719.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021719.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021719.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021721.hg.1 | TC09001692.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09021721.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021721.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021720.hg.1 | TC09001692.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09021720.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021720.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021720.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181181.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181181.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181181.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181181.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181181.hg.1 | TC0900011854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181180.hg.1 | TC0900011854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181179.hg.1 | TC0900011854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | ENST00000413714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011854.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181182.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181182.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181182.hg.1 | ENST00000339996.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181182.hg.1 | NM_001080483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181182.hg.1 | TC0900011854.hg.1 | Clariom D TC |
