| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1300007335.hg.1 | 
												NR_109839.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000422052.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000432995.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												ENST00000435636.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_046539.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_051993.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_051994.2 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												NR_109838.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1300149684.hg.1 | 
												TC1300007335.hg.1 | 
												Clariom D TC |