Assay Details
Gene Symbol: LINC00634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024355.1 | 1 - 2 | 232 | 69 | ||
GenBank mRNA | BC040153.1 | - | 1 - 2 | 121 | 69 | |
BC068568.1 | - | 1 - 2 | 232 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000349.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513922 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513929 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513928 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513926 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17286758 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11513923 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 3877 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 63513 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 119971 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295458 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3377187 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4526 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4525 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9946 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50647 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P03629726 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P03629726 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3377187 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295458 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3377187 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3295458 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200405 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513902 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513905 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513909 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513908 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513913 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513899 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200389 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513913 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513902 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513909 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513905 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513908 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200405 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200389 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513899 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513911 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03629726 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P420365 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513894 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513956 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513944 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513937 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513867 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513908 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286751 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513898 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513925 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286764 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513946 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513915 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513936 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513904 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286766 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513911 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513949 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513893 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513916 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513900 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513871 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513938 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513912 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513948 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513875 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513902 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513935 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513957 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286769 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286755 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513942 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286768 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513922 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286748 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513950 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513873 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286750 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513917 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513923 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286759 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286747 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513955 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513929 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513920 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513932 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513940 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513878 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513868 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11513951 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17286758 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03629729 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03629726 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8073547 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8073546 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006748.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006748.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006748.hg.1 | TC22000349.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22006749.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006749.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006749.hg.1 | TC22000349.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200153138.hg.1 | TC2200007504.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200153138.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153138.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153138.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153138.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | TC2200007504.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153131.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | TC2200007504.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153135.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153132.hg.1 | TC2200007504.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200153132.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153132.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153132.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153132.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153139.hg.1 | TC2200007504.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200153139.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153139.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153139.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153139.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | ENST00000505157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007504.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153137.hg.1 | TC2200007504.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200153137.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153137.hg.1 | NR_024355.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200153137.hg.1 | ENST00000381348.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200153137.hg.1 | NR_024355.1 | Refseq |
Human Genome U95 Set | 59594_PM_r_at | - | - |
Human Genome U95 Set | 59590_i_at | - | - |
Human Genome U95 Set | 59590_PM_i_at | - | - |
Human Genome U95 Set | 59594_r_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020512 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020512 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 524059 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1798043 | NR_024355.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1798043 | BC040153.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1798043 | BC068568.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1695799 | NR_024355.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1695799 | BC040153.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1695799 | BC068568.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1798043 | NR_024355.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1798043 | BC040153.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1798043 | BC068568.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1798043 | NR_024355.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1798043 | BC040153.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1798043 | BC068568.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1798043 | NR_024355.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1798043 | BC040153.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1798043 | BC068568.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695799 | NR_024355.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695799 | BC040153.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695799 | BC068568.1 | human_transcriptome |
