Assay Details
| Gene Symbol: ADGRE4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024075.1 | 1 - 2 | 77 | 74 | ||
| GenBank mRNA | AF489700.1 | - | 1 - 2 | 77 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001106.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015245.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015245.hg.1 | TC19001106.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015246.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015246.hg.1 | TC19001106.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900176108.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176108.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176108.hg.1 | TC1900009452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176108.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176108.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | TC1900009452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176106.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |

ADGRE4P
Genomic Map