Assay Details
Gene Symbol: CA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121632.1 | 1 - 2 | 121 | 76 | ||
NR_121633.1 | 1 - 2 | 121 | 76 | |||
GenBank mRNA | AW172333.1 | - | 2 - 3 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010993.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174750.hg.1 | TC0800010993.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000517697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000521761.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | ENST00000524052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121630.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121631.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121632.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | NR_121633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174749.hg.1 | TC0800010993.hg.1 | Clariom D TC |
