Assay Details
Gene Symbol: MOXD2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024346.3 | 5 - 6 | 851 | 150 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001936.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07030083.hg.1 | TC07001936.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07030083.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07030083.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700192686.hg.1 | TC0700012854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000460233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | ENST00000477615.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012854.hg.1 | NR_024346.3 | Refseq |
