Assay Details
Gene Symbol: B3GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005246931.3 | XP_005246988.1 | 1 - 2 | 276 | 116 | |
XM_011512085.2 | XP_011510387.1 | 1 - 2 | 276 | 116 | ||
GenBank mRNA | E07739.1 | - | 1 - 2 | 166 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005246931.3, XM_011512085.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
