Assay Details
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142641.1 | NP_001136113.1 | 7 - 8 | 1008 | 133 | |
XM_005266171.3 | XP_005266228.1 | 7 - 8 | 1119 | 133 | ||
XM_005266173.3 | XP_005266230.1 | 7 - 8 | 1120 | 133 | ||
XM_005266175.3 | XP_005266232.1 | 7 - 8 | 1122 | 133 | ||
XM_005266176.3 | XP_005266233.1 | 7 - 8 | 1118 | 133 | ||
XM_005266177.3 | XP_005266234.1 | 7 - 8 | 1114 | 133 | ||
XM_011534803.2 | XP_011533105.1 | 7 - 8 | 1121 | 133 | ||
XM_011534805.2 | XP_011533107.1 | 7 - 8 | 1121 | 133 | ||
XM_011534806.2 | XP_011533108.1 | 7 - 8 | 1124 | 133 | ||
XM_011534807.2 | XP_011533109.1 | 7 - 8 | 1120 | 133 | ||
XM_011534808.2 | XP_011533110.1 | 7 - 8 | 1116 | 133 | ||
XM_011534809.2 | XP_011533111.1 | 7 - 8 | 1119 | 133 | ||
XM_011534810.2 | XP_011533112.1 | 7 - 8 | 1121 | 133 | ||
XM_011534812.2 | XP_011533114.1 | 8 - 9 | 943 | 133 | ||
XM_011534816.2 | XP_011533118.1 | 7 - 8 | 1120 | 133 | ||
GenBank mRNA | AB046765.2 | - | 7 - 8 | 779 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001060.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12013992.hg.1 | ENST00000261673.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013992.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013992.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12013992.hg.1 | TC12001060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170875.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170875.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170875.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170875.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170875.hg.1 | TC1200009489.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200170876.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170876.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170876.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170876.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170876.hg.1 | TC1200009489.hg.1 | Clariom D TC |
