Assay Details
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142641.1 | NP_001136113.1 | 11 - 12 | 1587 | 68 | |
GenBank mRNA | AB046765.2 | - | 11 - 12 | 1358 | 68 | |
AF490258.1 | - | 4 - 5 | 453 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001060.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12013996.hg.1 | ENST00000261673.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013996.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013996.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12013996.hg.1 | TC12001060.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170884.hg.1 | TC1200009489.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170887.hg.1 | TC1200009489.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200170888.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170888.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170888.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170888.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170888.hg.1 | TC1200009489.hg.1 | Clariom D TC |
