Assay Details
| Gene Symbol: FLJ40194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034161.1 | 1 - 2 | 55 | 81 | ||
| GenBank mRNA | AK097513.1 | - | 1 - 2 | 81 | ||
| BI463223.1 | - | 1 - 2 | 60 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000641.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK097513.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352957 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352962 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352981 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930666 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16993731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353002 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352986 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352937 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352948 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352977 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352942 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352952 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353003 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930674 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30930707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10352941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10353007 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P42379 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 664 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 10614 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52384 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P418152 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P42379 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P42379 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 179457 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 86077 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P20887881 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03254896 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03254899 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P130491 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8008196 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8008197 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8008199 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8008198 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8008200 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8008195 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008000.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008000.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008000.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008000.hg.1 | TC17000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008004.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008004.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008004.hg.1 | TC17000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007999.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007999.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007999.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007999.hg.1 | TC17000641.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700160659.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160659.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160659.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160659.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160659.hg.1 | TC1700008196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160644.hg.1 | TC1700008196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160645.hg.1 | TC1700008196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160646.hg.1 | TC1700008196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700160658.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160658.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160658.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160658.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160658.hg.1 | TC1700008196.hg.1 | Clariom D TC |
| Human Genome U95 Set | 59607_f_at | - | - |
| Human Genome U95 Set | 59607_PM_f_at | - | - |
| Human Genome U95 Set | 78915_at | - | - |
| Human Genome U95 Set | 78915_PM_at | - | - |
| Human Genome U95 Set | 67378_at | - | - |
| Human Genome U95 Set | 67378_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009675 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 233123 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1800914 | AK097513.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1800914 | AK097513.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1800914 | AK097513.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1800914 | AK097513.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1800914 | AK097513.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1800914 | AK097513.1 | human_transcriptome |

FLJ40194
Genomic Map