Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005336.hg.1 |
ENST00000549830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005336.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005336.hg.1 |
TC12000417.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154632.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154632.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154632.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154632.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154632.hg.1 |
TC1200007658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200154636.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154636.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154636.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154636.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154636.hg.1 |
TC1200007658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
ENST00000549830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007658.hg.1 |
NR_027358.2 |
Refseq |