Assay Details
| Gene Symbol: CERS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256126.1 | NP_001243055.1 | 3 - 4 | 601 | 141 | |
| NM_203463.2 | NP_982288.1 | 3 - 4 | 601 | 141 | ||
| GenBank mRNA | AK172775.1 | - | 3 - 4 | 601 | 141 | |
| BC109284.1 | - | 3 - 4 | 451 | 40031838 | 141 | |
| BC109285.1 | - | 3 - 4 | 451 | 40031840 | 141 | |
| BE745491.1 | - | 118 | 141 | |||
| BX393696.2 | - | 3 - 4 | 531 | 141 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000996.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02014536.hg.1 | TC02000996.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014536.hg.1 | ENST00000305747.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014536.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014536.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014536.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169068.hg.1 | TC0200009895.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |

CERS6
Genomic Map