Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019452.hg.1 |
TC17002918.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019452.hg.1 |
ENST00000355612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019452.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012400.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
ENST00000355612.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
ENST00000578078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
NM_213656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206291.hg.1 |
TC1700012400.hg.1 |
Clariom D TC |