Assay Details
Gene Symbol: KRT39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213656.3 | NP_998821.3 | 5 - 6 | 1031 | 73 | |
GenBank mRNA | AJ786657.1 | - | 5 - 6 | 1031 | 73 | |
BC157887.1 | - | 5 - 6 | 1025 | 8862708 | 73 | |
BC171907.1 | - | 6 - 7 | 1069 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17002918.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 496238 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 687009 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 53237 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3268234 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3647 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3646 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25614 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3268234 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3268234 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_213656_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019618 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015149 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015146 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015145 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015148 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015147 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019615 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015150 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019619 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8015151 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019620 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019621 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019617 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8019616 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8015144 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8019614 | - | - |
Affymetrix Human Genome U219 Array | 11755944_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11755944_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR17019447.hg.1 | TC17002918.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17019447.hg.1 | ENST00000355612.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019447.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019449.hg.1 | TC17002918.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17019449.hg.1 | ENST00000355612.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019449.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019450.hg.1 | TC17002918.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17019450.hg.1 | ENST00000355612.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019450.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019448.hg.1 | TC17002918.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17019448.hg.1 | ENST00000355612.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019448.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206286.hg.1 | TC1700012400.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206288.hg.1 | TC1700012400.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700206284.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206284.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206284.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206284.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206284.hg.1 | TC1700012400.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206287.hg.1 | TC1700012400.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700206285.hg.1 | TC1700012400.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008300 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008300 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 187003 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2114137 | BC157887.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2114137 | BC157887.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2114137 | BC157887.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2114137 | BC157887.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_9779-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 290047 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_9779 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2114137 | BC157887.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2114137 | BC157887.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2114137 | BC157887.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2114137 | AJ786657.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1727626 | AJ786657.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1727626 | AJ786657.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2114137 | BC157887.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2114137 | AJ786657.1 | human_transcriptome |
