Assay Details
Gene Symbol: LIFR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103554.1 | 2 - 3 | 2542 | 98 | ||
GenBank mRNA | BC045578.1 | - | 2 - 3 | 498 | 98 | |
BG771462.1 | - | 2 - 3 | 503 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000514291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000514291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | ENST00000514291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148693.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148693.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148693.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148693.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148693.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500148694.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148694.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148694.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148694.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148694.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500733.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000514291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000514291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | ENST00000514291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103553.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007194.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148692.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148692.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148692.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148692.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148692.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500148696.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148696.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148696.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148696.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148696.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500148690.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148690.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148690.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148690.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148690.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500148691.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148691.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148691.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148691.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148691.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500148695.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148695.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148695.hg.1 | ENST00000500817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148695.hg.1 | NR_103554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148695.hg.1 | TC0500007194.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 234219 | - | - |
