Assay Details
| Gene Symbol: LOC101927482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110225.1 | 1 - 2 | 141 | 97 | ||
| GenBank mRNA | BF573965.1 | - | 139 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000605907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000605907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | ENST00000605907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000605907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000605907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | ENST00000605907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010337.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173005.hg.1 | TC0200010337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200173008.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173008.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173008.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173008.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173008.hg.1 | TC0200010337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | ENST00000430904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | ENST00000433933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | NR_110225.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | NR_110226.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200173006.hg.1 | TC0200010337.hg.1 | Clariom D TC |

LOC101927482
Genomic Map