Assay Details
| Gene Symbol: NFIA-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110617.1 | 5 - 6 | 811 | 82 | ||
| GenBank mRNA | BC030753.2 | - | 5 - 6 | 811 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | ENST00000418662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | ENST00000438559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | NR_110617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100207691.hg.1 | TC0100014377.hg.1 | Clariom D TC |

NFIA-AS2
Genomic Map