Assay Details
Gene Symbol: NFIA-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110617.1 | 6 - 7 | 884 | 81 | ||
GenBank mRNA | BC030753.2 | - | 6 - 7 | 884 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000421455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014377.hg.1 | NR_110618.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207689.hg.1 | TC0100014377.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | ENST00000418662.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207690.hg.1 | TC0100014377.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | ENST00000438559.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | NR_110617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207688.hg.1 | TC0100014377.hg.1 | Clariom D TC |
