Assay Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142300.1 | NP_001135772.1 | 3 - 4 | 691 | 116 | |
GenBank mRNA | BC067253.1 | - | 3 - 4 | 478 | 6500304 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001232.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000295414.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000339882.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000392209.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000420822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000452312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000468768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000468768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000468768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000468768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | ENST00000581168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_001142300.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NM_152523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010615.hg.1 | NR_039934.1 | Refseq |
