Assay Details
Gene Symbol: CATSPER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198137.1 | NP_937770.1 | 3 - 4 | 464 | 81 | |
XM_011541432.2 | XP_011539734.1 | 3 - 4 | 571 | 81 | ||
XM_011541433.2 | XP_011539735.1 | 3 - 4 | 1130 | 81 | ||
GenBank mRNA | BC128138.1 | - | 3 - 4 | 463 | 40112485 | 81 |
BC128139.1 | - | 3 - 4 | 464 | 40112493 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000337.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
neuronal action potential
sperm motility
response to progesterone
regulation of ion transmembrane transport
sperm-egg recognition
sodium ion transmembrane transport
sperm capacitation
regulation of postsynaptic membrane potential
calcium ion transmembrane transport
membrane depolarization during action potential
calcium activated cation channel activity
voltage-gated calcium channel activity
voltage-gated sodium channel activity
multicellular organism development
neuronal action potential
sperm motility
response to progesterone
regulation of ion transmembrane transport
sperm-egg recognition
sodium ion transmembrane transport
sperm capacitation
regulation of postsynaptic membrane potential
calcium ion transmembrane transport
membrane depolarization during action potential
calcium activated cation channel activity
voltage-gated calcium channel activity
voltage-gated sodium channel activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01005166.hg.1 | TC01000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005166.hg.1 | ENST00000456354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005166.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005164.hg.1 | TC01000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005164.hg.1 | ENST00000456354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005164.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005167.hg.1 | TC01000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005167.hg.1 | ENST00000456354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005167.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005168.hg.1 | TC01000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005168.hg.1 | ENST00000456354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005168.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005169.hg.1 | TC01000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005169.hg.1 | ENST00000456354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005169.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151937.hg.1 | TC0100007445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151939.hg.1 | TC0100007445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151938.hg.1 | TC0100007445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151936.hg.1 | TC0100007445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
