Assay Details
Gene Symbol: C12orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242696.1 | NP_001229625.1 | 2 - 3 | 563 | 83 | |
GenBank mRNA | BC038743.1 | - | 2 - 3 | 531 | 83 | |
BC117348.1 | - | 2 - 3 | 413 | 83 | ||
BC126393.1 | - | 2 - 3 | 413 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001514.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019889.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019889.hg.1 | TC12001514.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12019886.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019886.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019886.hg.1 | TC12001514.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12019887.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019887.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019887.hg.1 | TC12001514.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12019888.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019888.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019888.hg.1 | TC12001514.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181811.hg.1 | TC1200010741.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181810.hg.1 | TC1200010741.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181814.hg.1 | TC1200010741.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200181816.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181816.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181816.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181816.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181816.hg.1 | TC1200010741.hg.1 | Clariom D TC |
