Assay Details
Gene Symbol: AQP12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011511672.2 | XP_011509974.1 | 2 - 3 | 810 | 94 | |
GenBank mRNA | BC041460.1 | - | 3 - 4 | 505 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000407834.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000413999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000414322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000452886.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000459806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | ENST00000621682.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016330.hg.1 | NM_001102467.1 | Refseq |
