Assay Details
| Gene Symbol: CHRM3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832284.1 | - | 3 - 4 | 438 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1139 | LINC01139 | -1.2mb | Sense | Chr. 1 - 238480384 - 238486017 on NCBI Build GRCh38 | |
| Downstream | CHRM3 antisense RNA 1 | CHRM3-AS1 | 190.3kb | Sense | Chr. 1 - 239898019 - 239899872 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000427859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000428176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000458325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000593855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000596999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000600831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000610890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000627761.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | ENST00000628573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017904.hg.1 | NR_103776.1 | Refseq |

CHRM3-AS2
Genomic Map