Assay Details
Gene Symbol: FAAHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045483.1 | 6 - 7 | 506 | 140 | ||
GenBank mRNA | BI756183.1 | - | 3 - 4 | 197 | 140 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000586.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3414113 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35321 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3414113 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3414113 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01010024.hg.1 | TC01000586.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010024.hg.1 | ENST00000429784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010024.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010022.hg.1 | TC01000586.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010022.hg.1 | ENST00000429784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010022.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010023.hg.1 | TC01000586.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010023.hg.1 | ENST00000429784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010023.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158429.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158434.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158436.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158436.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158436.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158435.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158435.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158435.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158435.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158435.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158431.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Human Genome U95 Set | 47691_PM_r_at | - | - |
Human Genome U95 Set | 47691_r_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 169431 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 361497 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3304917 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_3304917 | - | - |
