Assay Details
Gene Symbol: SLC2A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033967.1 | 1 - 2 | 526 | 65 | ||
GenBank mRNA | AK056786.1 | - | 1 - 2 | 471 | 65 | |
BU657382.1 | - | 1 - 2 | 533 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000537.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165618 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165602 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20190320 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165613 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165616 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20190342 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165608 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165603 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165610 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00165599 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 150821 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 432729 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 496269 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 698481 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 230790 | XM_926717.1 | refseq_mrna_human_XM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 230790 | XM_938866.1 | refseq_mrna_human_XM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 855099 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 933850 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 907568 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3694746 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51036 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3694746 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P15100151 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15100136 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15100151 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 25174 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 67249 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10100498 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15100136 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15100151 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15100148 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00054343 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10094814 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35106179 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P17544041 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35106146 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15100136 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15100151 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1558832_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01008496.hg.1 | TC01000537.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01008496.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01008496.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01008494.hg.1 | TC01000537.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01008494.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01008497.hg.1 | TC01000537.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01008497.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01008497.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01008499.hg.1 | TC01000537.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01008499.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01008495.hg.1 | TC01000537.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01008495.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01008495.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156714.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156709.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156718.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156716.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156720.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156712.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008044.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | ENST00000416689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | ENST00000431759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | NR_033967.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100156711.hg.1 | TC0100008044.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1558832_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1558832_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558832_at2 | - | - |
Human Genome U95 Set | 75313_at | - | - |
Human Genome U95 Set | 75313_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021104 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021104 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 150796 | - | - |
