Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278505.1 | NP_001265434.1 | 3 - 4 | 317 | 144 | |
| NM_001322249.1 | NP_001309178.1 | 4 - 5 | 544 | 144 | ||
| NM_174931.3 | NP_777591.3 | 5 - 6 | 586 | 144 | ||
| XM_017003748.1 | XP_016859237.1 | 5 - 6 | 487 | 144 | ||
| GenBank mRNA | AK095667.1 | - | 3 - 4 | 250 | 144 | |
| AK289504.1 | - | 3 - 4 | 232 | 144 | ||
| AK294697.1 | - | 5 - 6 | 500 | 144 | ||
| BC071798.1 | - | 3 - 4 | 249 | 144 | ||
| BM285380.1 | - | 5 - 6 | 586 | 144 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000233.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02003453.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003453.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003453.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003450.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003450.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003450.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003450.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003452.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003452.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003452.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003452.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003456.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003456.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003456.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003456.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003455.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003455.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003455.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003455.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003449.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003449.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003449.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003454.hg.1 | TC02000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003454.hg.1 | ENST00000409774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003454.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150163.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150163.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150163.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150163.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150163.hg.1 | TC0200007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200150162.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150162.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150162.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150162.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150162.hg.1 | TC0200007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150165.hg.1 | TC0200007249.hg.1 | Clariom D TC |

GPATCH11
Genomic Map