Assay Details
| Gene Symbol: FER1L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039112.2 | NP_001034201.2 | 11 - 12 | 1614 | 99 | |
| XM_006716618.3 | XP_006716681.1 | 11 - 12 | 1541 | 99 | ||
| XM_011517231.2 | XP_011515533.1 | 12 - 13 | 1580 | 99 | ||
| XM_011517232.2 | XP_011515534.1 | 12 - 13 | 1721 | 99 | ||
| XM_011517233.2 | XP_011515535.1 | 11 - 12 | 1682 | 99 | ||
| XM_011517235.2 | XP_011515537.1 | 12 - 13 | 1581 | 99 | ||
| GenBank mRNA | AB196633.1 | - | 11 - 12 | 1614 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000721.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000399018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | ENST00000522917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008759.hg.1 | NM_001039112.2 | Refseq |

FER1L6
Genomic Map