Assay Details
| Gene Symbol: LINC01423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110545.1 | 2 - 3 | 239 | 101 | ||
| GenBank mRNA | AI752367.1 | - | 1 - 2 | 76 | 101 | |
| DA464432.1 | - | 2 - 3 | 239 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | TC2100008141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154839.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008141.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | TC2100008141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154838.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | NR_110545.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | TC2100008141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | ENST00000414189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | ENST00000436845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100154840.hg.1 | NR_110545.1 | Refseq |

LINC01423
Genomic Map