Assay Details
| Gene Symbol: MFSD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080473.1 | NP_001073942.1 | 11 - 12 | 1190 | 68 | |
| XM_005264301.4 | XP_005264358.1 | 11 - 12 | 2251 | 68 | ||
| XM_017004063.1 | XP_016859552.1 | 10 - 11 | 2344 | 68 | ||
| XM_017004064.1 | XP_016859553.1 | 10 - 11 | 2329 | 68 | ||
| XM_017004065.1 | XP_016859554.1 | 9 - 10 | 2260 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004951.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001433.hg.1 | TC02004951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001433.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001433.hg.1 | ENST00000406420.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001433.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | ENST00000495018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016412.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | ENST00000338315.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | ENST00000406420.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | ENST00000469562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | NM_001080473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221108.hg.1 | TC0200016412.hg.1 | Clariom D TC |

MFSD2B
Genomic Map