Assay Details
Gene Symbol: LOC284865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038460.1 | 3 - 4 | 1708 | 67 | ||
GenBank mRNA | AK092552.1 | - | 3 - 4 | 1708 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000507.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432722 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432746 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432752 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103511 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432807 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432797 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432745 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432735 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103529 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432784 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17256485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432743 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432796 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432724 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432812 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432785 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432776 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432769 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432756 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432727 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17256483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432728 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432781 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432768 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103548 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432788 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432787 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103475 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432805 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432794 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432792 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103523 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432795 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432740 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432793 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432757 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432770 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432766 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103531 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432736 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432819 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103455 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432721 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432774 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432723 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432799 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432801 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432715 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432720 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32103499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432772 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432806 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432719 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432726 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432810 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432791 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11432790 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 700741 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 477486 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 576834 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 285417 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 223787 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03601982 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13959179 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03601982 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03601978 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03601988 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03601988 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03601982 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1564109_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR22009404.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009404.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009404.hg.1 | TC22000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009407.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009407.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009407.hg.1 | TC22000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009405.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009405.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009405.hg.1 | TC22000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009406.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009406.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009406.hg.1 | TC22000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157355.hg.1 | TC2200008025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200157355.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157355.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157355.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157355.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157358.hg.1 | TC2200008025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200157358.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157358.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157358.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157358.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157356.hg.1 | TC2200008025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200157356.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157356.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157356.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157356.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157357.hg.1 | TC2200008025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200157357.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157357.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200157357.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200157357.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | ENST00000506039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008025.hg.1 | NR_038460.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1564109_at | - | - |
Human Genome U133 Plus 2.0 Array | 1564109_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564109_at4 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019484 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019484 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 104102 | - | - |
