Assay Details
Gene Symbol: LOC101927438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110224.1 | 2 - 3 | 331 | 118 | ||
GenBank mRNA | AK097952.1 | - | 2 - 3 | 326 | 118 | |
AV712717.1 | - | 2 - 3 | 330 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
