Assay Details
Gene Symbol: LOC101927286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120324.1 | 4 - 5 | 747 | 85 | ||
GenBank mRNA | BC038379.1 | - | 4 - 5 | 747 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spermatogenesis associated 8 | SPATA8 | SRG8 | -584.8kb | Anti-sense | Chr. 15 - 96783407 - 96785615 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101927310 | LOC101927310 | 155.5kb | Sense | Chr. 15 - 97553284 - 97560698 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
