Assay Details
| Gene Symbol: LOC101929679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120366.1 | 4 - 5 | 548 | 114 | ||
| GenBank mRNA | AL357200.1 | - | 1 - 2 | 410 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | A-kinase anchoring protein 13 | AKAP13 |
AKAP-13, AKAP-Lbc, ARHGEF13, BRX, HA-3, Ht31, LBC, PRKA13, PROTO-LB, PROTO-LBC, c-lbc, p47 |
-6.2kb | Anti-sense | Chr. 15 - 85380616 - 85749358 on NCBI Build GRCh38 |
| Downstream | kelch like family member 25 | KLHL25 |
ENC-2, ENC2 |
3.4kb | Sense | Chr. 15 - 85759326 - 85794958 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929679
Genomic Map