Assay Details
Gene Symbol: LINC00996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034033.1 | 3 - 4 | 982 | 81 | ||
GenBank mRNA | AK097158.1 | - | 3 - 4 | 884 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001001.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867665 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867648 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132937 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867626 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132956 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132952 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16037745 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16037765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867622 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867628 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867617 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132942 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132890 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867616 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867657 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867632 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132931 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867658 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867618 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867635 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132938 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16037762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867623 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867634 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867652 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132934 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867630 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16037753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05867667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26132951 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 26111 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 899823 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 893097 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 942351 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 608693 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3714477 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17753 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P38252845 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P38252867 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3714477 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3714477 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01834741 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P38252845 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 140006 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P25844122 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38252867 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01834743 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01834739 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01834741 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38252845 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P539615 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38252845 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38252867 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01834741 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1560762_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR07015244.hg.1 | TC07001001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015244.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07015244.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168706.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168706.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168706.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168706.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168706.hg.1 | TC0700009674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168705.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168705.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168705.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168705.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168705.hg.1 | TC0700009674.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1560762_at | - | - |
Human Genome U133 Plus 2.0 Array | 1560762_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560762_at4 | - | - |
Human Genome U95 Set | 44891_PM_r_at | - | - |
Human Genome U95 Set | 44891_r_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035315 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035315 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 696902 | - | - |
