Assay Details
Gene Symbol: LOC100271832 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027097.1 | 2 - 3 | 316 | 95 | ||
GenBank mRNA | BC035107.1 | - | 2 - 3 | 321 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 486 | LINC00486 | Sense | Chr. 2 - 32825443 - 32937203 on NCBI Build GRCh38 | ||
Downstream | latent transforming growth factor beta binding protein 1 | LTBP1 | 18.9kb | Sense | Chr. 2 - 32947129 - 33399509 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000218.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02003213.hg.1 | TC02000218.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003213.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003213.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003215.hg.1 | TC02000218.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003215.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003215.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003214.hg.1 | TC02000218.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003214.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003214.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149722.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149722.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149722.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149722.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149722.hg.1 | TC0200007199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149727.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149727.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149727.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149727.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149727.hg.1 | TC0200007199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200149726.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149726.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149726.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149726.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149726.hg.1 | TC0200007199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200149725.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149725.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149725.hg.1 | ENST00000440786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149725.hg.1 | NR_027097.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149725.hg.1 | TC0200007199.hg.1 | Clariom D TC |
