Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184658.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184658.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184658.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184658.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184658.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184662.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
ENST00000436187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184661.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200184663.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184663.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184663.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184663.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184663.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200184659.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184659.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184659.hg.1 |
ENST00000454224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184659.hg.1 |
NR_110257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184659.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |