Assay Details
Gene Symbol: TTBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032538.1 | NP_115927.1 | 2 - 3 | 381 | 79 | |
XM_011514947.2 | XP_011513249.1 | 1 - 2 | 171 | 79 | ||
XM_017011364.1 | XP_016866853.1 | 1 - 2 | 171 | 79 | ||
XM_017011365.1 | XP_016866854.1 | 1 - 2 | 171 | 79 | ||
XM_017011366.1 | XP_016866855.1 | 1 - 2 | 171 | 79 | ||
XM_017011367.1 | XP_016866856.1 | 1 - 2 | 171 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000598.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06007046.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007046.hg.1 | ENST00000259750.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007046.hg.1 | ENST00000393984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007046.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007047.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007047.hg.1 | ENST00000259750.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007047.hg.1 | ENST00000393984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007047.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007048.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007048.hg.1 | ENST00000259750.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007048.hg.1 | ENST00000393984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007048.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007050.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007050.hg.1 | ENST00000259750.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007050.hg.1 | ENST00000304139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007050.hg.1 | ENST00000393984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007050.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007045.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007045.hg.1 | ENST00000393984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007045.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156364.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156364.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156364.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156364.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156364.hg.1 | TC0600008075.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600156363.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156363.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156363.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156363.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156363.hg.1 | TC0600008075.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156367.hg.1 | TC0600008075.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600156365.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156365.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156365.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156365.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156365.hg.1 | TC0600008075.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156362.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156362.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156362.hg.1 | TC0600008075.hg.1 | Clariom D TC |
