Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007052.hg.1 |
TC06000598.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007052.hg.1 |
ENST00000259750.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007052.hg.1 |
ENST00000304139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007052.hg.1 |
ENST00000393984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007052.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008075.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
ENST00000259750.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
ENST00000304139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
NM_032538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156369.hg.1 |
TC0600008075.hg.1 |
Clariom D TC |