Assay Details
Gene Symbol: LOC102724651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110798.1 | 3 - 4 | 272 | 110 | ||
GenBank mRNA | BX088865.1 | - | 3 - 4 | 236 | 110 | |
DB079133.1 | - | 3 - 4 | 273 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000579506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000579506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | TC1800007345.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150320.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | TC1800007345.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150321.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000578610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000579506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000579506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | ENST00000581198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007345.hg.1 | NR_110798.1 | Refseq |
