| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050728.hg.1 |
TC01003289.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050728.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050728.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050730.hg.1 |
TC01003289.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050730.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050730.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050729.hg.1 |
TC01003289.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050729.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050729.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219732.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219732.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219732.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219732.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219732.hg.1 |
TC0100015901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219730.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219730.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219730.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219730.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219730.hg.1 |
TC0100015901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219735.hg.1 |
TC0100015901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219731.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219731.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219731.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219731.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219731.hg.1 |
TC0100015901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219733.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219733.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219733.hg.1 |
TC0100015901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |