Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015901.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219729.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219729.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219729.hg.1 |
ENST00000368525.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219729.hg.1 |
NM_001010979.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219729.hg.1 |
TC0100015901.hg.1 |
Clariom D TC |