Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008724.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008724.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008724.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008724.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800159847.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800159847.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800159847.hg.1 |
TC1800008724.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800159847.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800159847.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800159848.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800159848.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800159848.hg.1 |
TC1800008724.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800159848.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800159848.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008724.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008724.hg.1 |
ENST00000565170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008724.hg.1 |
NR_110800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008724.hg.1 |
NR_110800.1 |
Refseq |