Assay Details
| Gene Symbol: LOC101927200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096842.1 | - | 1 - 2 | 102 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gametocyte specific factor 1 like | GTSF1L |
C20orf65, FAM112A, dJ1028D15.4 |
-62.9kb | Anti-sense | Chr. 20 - 43726161 - 43727135 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105372626 | LOC105372626 | 82.4kb | Sense | Chr. 20 - 43894718 - 43895468 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927200
Genomic Map