| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166221.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166221.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166221.hg.1 | 
												TC2000009463.hg.1 | 
												Clariom D TC | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166221.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166221.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166222.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166222.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166222.hg.1 | 
												TC2000009463.hg.1 | 
												Clariom D TC | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166222.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR2000166222.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												ENST00000438154.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC2000009463.hg.1 | 
												NR_110016.1 | 
												Refseq |